APO415184499 Apollo Scientific Ltd

2,6-Difluoro-3-(trifluoromethyl)benzonitrile

  • MDL number MFCD27937181
  • Cas Number : 1806333-92-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Physical Form Liquid
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Product Link https://store.apolloscientific.co.uk/product/26-difluoro-3-trifluoromethylbenzonitrile

Safety Information

signalword Danger
pcodes P260;P280
MSDS https://store.apolloscientific.co.uk/product/26-difluoro-3-trifluoromethylbenzonitrile
pictograms GHS06
hstatements H301;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart