APO415184503 Apollo Scientific Ltd

2,4-Difluoro-5-hydroxybenzotrifluoride

  • MDL number MFCD27937185
  • Cas Number : 1632283-20-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 2,4-Difluoro-5-(trifluoromethyl)phenol
Storage temp. Ambient Temperature
Physical Form Liquid-Oil
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=60564

Safety Information

signalword Danger
pcodes P260;P271;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=60564
pictograms GHS05;GHS07
hstatements H302+H312+H332;H314
Availability details for
Quantity
  • Availability
  • Add to Cart