APO415184518 Apollo Scientific Ltd

5-Bromo-2-chloro-4-fluorophenyl trifluoromethanesulphonate

  • MDL number MFCD27937161
  • Cas Number : 1820619-90-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 5-Bromo-2-chloro-4-fluorophenyl triflate
Physical Form Liquid
Shipping Temperature Ship at ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=60665

Safety Information

signalword Danger
msds http://www.apolloscientific.co.uk/display_item.php?id=60665
pictograms GHS05
Availability details for
Quantity
  • Availability
  • Add to Cart