APO415184537 Apollo Scientific Ltd

1-(3-Chloropyridin-2-yl)ethanone

  • MDL number MFCD11656222
  • Cas Number : 131109-75-6
  • Molecular Weight : 155.58

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 1-(3-chloro-2-pyridinyl)ethanone
Inchi Code 1S/C7H6ClNO/c1-5(10)7-6(8)3-2-4-9-7/h2-4H,1H3
Inchi Key GMXZSVFWDQBSFR-UHFFFAOYSA-N

Description

Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/1-3-chloropyridin-2-ylethanone

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/1-3-chloropyridin-2-ylethanone
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart