APO415184595 Apollo Scientific Ltd

2-(tert-Butoxy)pyridine-3-boronic acid

  • MDL number MFCD26397235
  • Cas Number : 1245898-82-1
  • Molecular Weight : 195.03

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name (2-(tert-butoxy)pyridin-3-yl)boronic acid
Inchi Code 1S/C9H14BNO3/c1-9(2,3)14-8-7(10(12)13)5-4-6-11-8/h4-6,12-13H,1-3H3
Inchi Key HNPJPQRHQRFFOW-UHFFFAOYSA-N

Description

Synonym 3-Borono-2-(tert-butoxy)pyridine
Storage temp. Store at 2-8
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/2-tert-butoxypyridine-3-boronic-acid

Safety Information

signalword Warning
pcodes P260;P280
MSDS https://store.apolloscientific.co.uk/product/2-tert-butoxypyridine-3-boronic-acid
pictograms GHS07
hstatements H302+H312+H332;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart