APO415184810 Apollo Scientific Ltd

Thiazolo[5,4-c]pyridine-2-thiol

  • MDL number MFCD12755950
  • Cas Number : 116990-44-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link https://store.apolloscientific.co.uk/product/thiazolo54-cpyridine-2-thiol

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/thiazolo54-cpyridine-2-thiol
pictograms GHS07
hstatements H302+H312+H332;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart