APO415184834 Apollo Scientific Ltd

1-Phenylcyclopropan-1-amine

  • MDL number MFCD05863520
  • Cas Number : 41049-53-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link https://store.apolloscientific.co.uk/product/1-phenylcyclopropan-1-amine

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/1-phenylcyclopropan-1-amine
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart