APO415184913 Apollo Scientific Ltd

4-Chloro-1H-imidazo[4,5-c]pyridine

  • MDL number MFCD08689582
  • Cas Number : 2770-01-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 264
Purity 98
Storage temp. Store at 2-8
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link https://store.apolloscientific.co.uk/product/4-chloro-1h-imidazo45-cpyridine

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/4-chloro-1h-imidazo45-cpyridine
pictograms GHS07
hstatements H302+H332;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart