APO415184966 Apollo Scientific Ltd

2-(4-Bromo-phenyl)-pyrrolidine

  • MDL number MFCD02663432
  • Cas Number : 383127-22-8
  • Molecular Weight : 226.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 2-(4-bromophenyl)pyrrolidine
Inchi Code 1S/C10H12BrN/c11-9-5-3-8(4-6-9)10-2-1-7-12-10/h3-6,10,12H,1-2,7H2
Inchi Key HIJZBROSVFKSCP-UHFFFAOYSA-N

Description

Purity 97%
Flash Point 287.2/760mm
Storage temp. Store at 2-8
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/2-4-bromo-phenyl-pyrrolidine

Safety Information

signalword Warning
pcodes P280;P301+P312;P302+P352
MSDS https://store.apolloscientific.co.uk/product/2-4-bromo-phenyl-pyrrolidine
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart