APO415184985 Apollo Scientific Ltd

2-Methoxy-6,7,8,9-tetrahydrobenzocyclohepten-5-one

  • MDL number MFCD07369971
  • Cas Number : 6500-65-8
  • Molecular Weight : 190.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 2-methoxy-6,7,8,9-tetrahydro-5H-benzo[a]cyclohepten-5-one
Inchi Code 1S/C12H14O2/c1-14-10-6-7-11-9(8-10)4-2-3-5-12(11)13/h6-8H,2-5H2,1H3
Inchi Key WLRMIOGKWITDNU-UHFFFAOYSA-N

Description

Synonym 2-Methoxy-6,7,8,9-tetrahydro-5H-benzo[7]annulen-5-one
Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/2-methoxy-6789-tetrahydrobenzocyclohepten-5-one

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/2-methoxy-6789-tetrahydrobenzocyclohepten-5-one
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart