APO415185000 Apollo Scientific Ltd

3-Chloro-5-methoxyaniline

  • MDL number MFCD11035922
  • Cas Number : 10272-06-7
  • Molecular Weight : 157.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 3-chloro-5-methoxyaniline
Inchi Code 1S/C7H8ClNO/c1-10-7-3-5(8)2-6(9)4-7/h2-4H,9H2,1H3
Inchi Key XLAVBRTYEPFKSD-UHFFFAOYSA-N

Description

Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/3-chloro-5-methoxyaniline

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/3-chloro-5-methoxyaniline
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart