APO415185030 Apollo Scientific Ltd

1-Fluorocyclopropane-1-carboxylic acid

  • MDL number MFCD18205954
  • Cas Number : 137081-41-5
  • Molecular Weight : 104.08

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

bp 72-76
Iupac Name 1-fluorocyclopropanecarboxylic acid
Inchi Code 1S/C4H5FO2/c5-4(1-2-4)3(6)7/h1-2H2,(H,6,7)
Inchi Key NDCPERCVXDYEFU-UHFFFAOYSA-N

Description

Purity 97%
Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 72-76
Product Link https://store.apolloscientific.co.uk/product/1-fluorocyclopropane-1-carboxylic-acid

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/1-fluorocyclopropane-1-carboxylic-acid
pictograms GHS07
hstatements H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart