APO415185055 Apollo Scientific Ltd

5-Bromo-4-fluoro-2-nitrotoluene

  • MDL number MFCD07777174
  • Cas Number : 948294-26-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 1-Bromo-2-fluoro-5-methyl-4-nitrobenzene, 4-Bromo-5-fluoro-2-methylnitrobenzene
Physical Form Solid
Shipping Temperature Ship at ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=60668

Safety Information

signalword Warning
msds http://www.apolloscientific.co.uk/display_item.php?id=60668
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart