APO415185070 Apollo Scientific Ltd

5-Bromo-6-fluoro-2-oxindole

  • MDL number MFCD26086914
  • Cas Number : 944805-66-7
  • Molecular Weight : 230.04

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 5-Bromo-6-fluoroindolin-2-one, 5-Bromo-1,3-dihydro-6-fluoro-2H-indol-2-one
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/5-bromo-6-fluoro-2-oxindole

Safety Information

signalword Warning
pcodes P260;P280
MSDS https://store.apolloscientific.co.uk/product/5-bromo-6-fluoro-2-oxindole
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart