APO415185078 Apollo Scientific Ltd

2-Amino-4-(difluoromethyl)pyridine

  • MDL number MFCD25476528
  • Cas Number : 1346536-47-7
  • Molecular Weight : 144.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 4-(difluoromethyl)pyridin-2-amine
Inchi Code 1S/C6H6F2N2/c7-6(8)4-1-2-10-5(9)3-4/h1-3,6H,(H2,9,10)
Inchi Key CXNARPJWYBRIKJ-UHFFFAOYSA-N

Description

Synonym 4-(Difluoromethyl)pyridin-2-amine
Purity 98%
Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/2-amino-4-difluoromethylpyridine

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/2-amino-4-difluoromethylpyridine
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart