APO415185209 Apollo Scientific Ltd

Dasatinib

  • MDL number MFCD11046566
  • Cas Number : 302962-49-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 283
Synonym N-(2-Chloro-6-methylphenyl)-2-((6-(4-(2-hydroxyethyl)piperazin-1-yl)-2-methylpyrimidin-4-yl)amino)thiazole-5-carboxamide
Storage temp. Store at -20
Physical Form Solid
Shipping Temperature Ship on Ice
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=61214

Safety Information

signalword Danger
pcodes P260;P271;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=61214
pictograms GHS06;GHS08
hstatements H301;H360D;H362
Availability details for
Quantity
  • Availability
  • Add to Cart