APO423213828 Apollo Scientific Ltd

3-Chloro-5-(trifluoromethoxy)phenacyl bromide

  • MDL number MFCD27975441
  • Cas Number : 1796558-59-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 2-Bromo-1-[3-chloro-5-(trifluoromethoxy)phenyl]ethanone
Physical Form Liquid
Shipping Temperature Ship at ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=61890

Safety Information

signalword Danger
pcodes P271;P260;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=61890
pictograms GHS05
hstatements H314
Availability details for
Quantity
  • Availability
  • Add to Cart