APO423213908 Apollo Scientific Ltd

8-Fluoro-3-iodoquinoline

  • MDL number MFCD07700291
  • Cas Number : 866782-59-4
  • Molecular Weight : 273.05

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 8-fluoro-3-iodoquinoline
Inchi Code 1S/C9H5FIN/c10-8-3-1-2-6-4-7(11)5-12-9(6)8/h1-5H
Inchi Key PFDRCQPNBZLTJQ-UHFFFAOYSA-N

Description

Purity 99%
Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/8-fluoro-3-iodoquinoline

Safety Information

signalword Warning
pcodes P271;P260;P280
MSDS https://store.apolloscientific.co.uk/product/8-fluoro-3-iodoquinoline
pictograms GHS07
hstatements H302+H312+H332;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart