APO423213919 Apollo Scientific Ltd

5-Iodo-2-nitrophenol

  • MDL number MFCD12923199
  • Cas Number : 27783-55-7
  • Molecular Weight : 265.01

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

bp 92-96
Iupac Name 5-iodo-2-nitrophenol
Inchi Code 1S/C6H4INO3/c7-4-1-2-5(8(10)11)6(9)3-4/h1-3,9H
Inchi Key NWDXJGPGCDHUJO-UHFFFAOYSA-N

Description

Synonym 2-Hydroxy-4-iodonitrobenzene
Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 92-96
Product Link https://store.apolloscientific.co.uk/product/5-iodo-2-nitrophenol

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/5-iodo-2-nitrophenol
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart