APO423213943 Apollo Scientific Ltd

2-Bromo-6-methoxybenzotrifluoride

  • MDL number MFCD13194484
  • Cas Number : 1214345-25-1
  • Molecular Weight : 255.03

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 1-Bromo-3-methoxy-2-(trifluoromethyl)benzene, 3-Bromo-2-(trifluoromethyl)phenyl methyl ether
Purity 95
Physical Form Liquid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/2-bromo-6-methoxybenzotrifluoride

Safety Information

signalword Warning
pcodes P271;P260;P280
MSDS https://store.apolloscientific.co.uk/product/2-bromo-6-methoxybenzotrifluoride
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart