APO423214057 Apollo Scientific Ltd

2-Bromo-3-methoxybenzaldehyde

  • MDL number MFCD09834663
  • Cas Number : 10401-18-0
  • Molecular Weight : 215.05

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

bp 67-69
Iupac Name 2-bromo-3-methoxybenzaldehyde
Inchi Code 1S/C8H7BrO2/c1-11-7-4-2-3-6(5-10)8(7)9/h2-5H,1H3
Inchi Key KELPUINOQGYPKR-UHFFFAOYSA-N

Description

Purity 95%
Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 67-69
Product Link https://store.apolloscientific.co.uk/product/2-bromo-3-methoxybenzaldehyde

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/2-bromo-3-methoxybenzaldehyde
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart