APO423214058 Apollo Scientific Ltd

Methyl 2-amino-6-fluoro-3-nitrobenzoate

  • MDL number MFCD09839221
  • Cas Number : 346691-23-4
  • Molecular Weight : 214.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 93-95
Synonym 3-Fluoro-2-(methoxycarbonyl)-6-nitroaniline
Physical Form Solid-Crystals
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/methyl-2-amino-6-fluoro-3-nitrobenzoate

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/methyl-2-amino-6-fluoro-3-nitrobenzoate
pictograms GHS07
hstatements H302+H312+H332;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart