APO423214083 Apollo Scientific Ltd

3-Bromo-2-iodophenol

  • MDL number MFCD08166320
  • Cas Number : 855836-52-1
  • Molecular Weight : 298.91

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/3-bromo-2-iodophenol

Safety Information

signalword Warning
pcodes P271;P260;P280
MSDS https://store.apolloscientific.co.uk/product/3-bromo-2-iodophenol
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart