APO423214084 Apollo Scientific Ltd

2-Bromo-6-iodoaniline

  • MDL number MFCD18391615
  • Cas Number : 84483-27-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Physical Form Solid
Shipping Temperature Ship at ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=61882

Safety Information

signalword Warning
msds http://www.apolloscientific.co.uk/display_item.php?id=61882
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart