APO423214103 Apollo Scientific Ltd

2-Chloro-6-fluoro-3-nitrotoluene

  • MDL number MFCD21603723
  • Cas Number : 90292-62-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 37-39
Synonym 2-Chloro-4-fluoro-3-methyl-1-nitrobenzene, 2-Chloro-4-fluoro-3-methylnitrobenzene
Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=61590

Safety Information

signalword Warning
pcodes P271;P260;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=61590
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart