APO423214180 Apollo Scientific Ltd

5-Iodo-2-nitroanisole

  • MDL number MFCD18397934
  • Cas Number : 860585-81-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 92
Synonym 4-Iodo-2-methoxynitrobenzene, 4-Iodo-2-methoxy-1-nitrobenzene, 5-Iodo-2-nitrophenyl methyl ether
Storage temp. Ambient temperature
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=61612

Safety Information

signalword Warning
pcodes P261;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=61612
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart