APO424014553 Apollo Scientific Ltd

2-Fluoro-4-hydroxy-5-nitrobenzaldehyde

  • MDL number MFCD27995610
  • Cas Number : 394-34-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Product Link https://store.apolloscientific.co.uk/product/2-fluoro-4-hydroxy-5-nitrobenzaldehyde-95

Safety Information

signalword Warning
pcodes P260;P280
MSDS https://store.apolloscientific.co.uk/product/2-fluoro-4-hydroxy-5-nitrobenzaldehyde-95
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart