APO424014571 Apollo Scientific Ltd

Methyl 2-bromo-6-iodobenzoate

  • MDL number MFCD11870176
  • Cas Number : 1261840-81-6
  • Molecular Weight : 340.94

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name methyl 2-bromo-6-iodobenzoate
Inchi Code 1S/C8H6BrIO2/c1-12-8(11)7-5(9)3-2-4-6(7)10/h2-4H,1H3
Inchi Key FYAVLNMQBCJMLV-UHFFFAOYSA-N

Description

Purity 95%
Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/methyl-2-bromo-6-iodobenzoate

Safety Information

signalword Danger
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/methyl-2-bromo-6-iodobenzoate
pictograms GHS06
hstatements H301;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart