APO424014636 Apollo Scientific Ltd

Spiro[indole-3,4'-piperidin]-2(1H)-one, N1'-BOC protected

  • MDL number MFCD10566051
  • Cas Number : 252882-60-3
  • Molecular Weight : 302.37

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym tert-Butyl 1,2-dihydro-2-oxo-1'H-spiro[indole-3,4'-piperidine]-1'-carboxylate, 1'-(tert-Butoxycarbonyl)-1,2-dihydro-2-oxospiro[indole-3,4'-piperidine]
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/spiroindole-34-piperidin-21h-one-n1-boc-protected

Safety Information

signalword Warning
pcodes P260;P280
MSDS https://store.apolloscientific.co.uk/product/spiroindole-34-piperidin-21h-one-n1-boc-protected
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart