APO424014671 Apollo Scientific Ltd

3-Fluoro-4-iodobenzotrifluoride

  • MDL number MFCD07771984
  • Cas Number : 132554-73-5
  • Molecular Weight : 290

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Physical Form Liquid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/3-fluoro-4-iodobenzotrifluoride

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/3-fluoro-4-iodobenzotrifluoride
pictograms GHS07
hstatements H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart