APO424014775 Apollo Scientific Ltd

2-Iodo-6-methoxybenzonitrile

  • MDL number MFCD18905184
  • Cas Number : 66195-38-8
  • Molecular Weight : 259.05

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 2-iodo-6-methoxybenzonitrile
Inchi Code 1S/C8H6INO/c1-11-8-4-2-3-7(9)6(8)5-10/h2-4H,1H3
Inchi Key JULARJRMSOOVTP-UHFFFAOYSA-N

Description

Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/2-iodo-6-methoxybenzonitrile

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/2-iodo-6-methoxybenzonitrile
pictograms GHS07
hstatements H302+H312+H332;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart