APO424014797 Apollo Scientific Ltd

4-Bromo-5-chloro-2-fluorobenzoic acid

  • MDL number MFCD19686184
  • Cas Number : 1349708-91-3
  • Molecular Weight : 253.45

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 152-154
Purity 98
Physical Form Solid-Powder
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/4-bromo-5-chloro-2-fluorobenzoic-acid

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/4-bromo-5-chloro-2-fluorobenzoic-acid
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart