APO424014879 Apollo Scientific Ltd

Octahydro-4H-isoindol-4-one, N-BOC protected

  • MDL number MFCD17170012
  • Cas Number : 879687-92-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym tert-Butyl 4-oxooctahydro-2H-isoindole-2-carboxylate, 2-(tert-Butoxycarbonyl)-4-oxooctahydro-2H-isoindole
Physical Form Solid
Shipping Temperature Ship at ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=62849

Safety Information

signalword Warning
msds http://www.apolloscientific.co.uk/display_item.php?id=62849
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart