APO424014936 Apollo Scientific Ltd

2-(Bromomethyl)phenyl acetate

  • MDL number MFCD20487652
  • Cas Number : 704-65-4
  • Molecular Weight : 229.07

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 2-(bromomethyl)phenyl acetate
Inchi Code 1S/C9H9BrO2/c1-7(11)12-9-5-3-2-4-8(9)6-10/h2-5H,6H2,1H3
Inchi Key RHZMCEFGRRLLAP-UHFFFAOYSA-N

Description

Storage temp. Store at -20
Physical Form Liquid-Oil
Shipping Temperature Shipped with Ice Packs
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/2-bromomethylphenyl-acetate

Safety Information

signalword Danger
pcodes P260;P280
MSDS https://store.apolloscientific.co.uk/product/2-bromomethylphenyl-acetate
pictograms GHS05
hstatements H314
Availability details for
Quantity
  • Availability
  • Add to Cart