APO424015051 Apollo Scientific Ltd

1-Benzyl-D-proline

  • MDL number MFCD06656733
  • Cas Number : 56080-99-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Physical Form Solid
Shipping Temperature Ship at ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=62573

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=62573
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart