APO424015054 Apollo Scientific Ltd

5-Bromobenzene-1,2,3-triol

  • MDL number MFCD22555992
  • Cas Number : 16492-75-4
  • Molecular Weight : 205.01

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Storage temp. Store at 2-8
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/5-bromobenzene-123-triol

Safety Information

signalword Warning
pcodes P271;P260;P280
MSDS https://store.apolloscientific.co.uk/product/5-bromobenzene-123-triol
pictograms GHS07
hstatements H302+H332;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart