APO424015116 Apollo Scientific Ltd

1-Oxa-7-azaspiro[4.5]decan-2-one hydrochloride

  • MDL number MFCD18642911
  • Cas Number : 1314961-56-2
  • Molecular Weight : 191.66

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 2-Oxo-1-oxa-7-azaspiro[4.5]decane hydrochloride
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/1-oxa-7-azaspiro45decan-2-one-hydrochloride

Safety Information

signalword Warning
pcodes P260;P280;P312
MSDS https://store.apolloscientific.co.uk/product/1-oxa-7-azaspiro45decan-2-one-hydrochloride
pictograms GHS07
hstatements H302+H312+H332;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart