APO424015195 Apollo Scientific Ltd

4-(5-{4-[(Pent-1-yl)oxy]phenyl}isoxazol-3-yl)benzoic acid

  • MDL number MFCD20267356
  • Cas Number : 179162-55-1
  • Molecular Weight : 351.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 4-{5-[4-(Pentyloxy)phenyl]-1,2-oxazol-3-yl}benzoic acid, 3-(4-Carboxyphenyl)-5-[4-(pentyloxy)phenyl]-1,2-oxazole
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/4-5-4-pent-1-yloxyphenylisoxazol-3-ylbenzoic-acid

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/4-5-4-pent-1-yloxyphenylisoxazol-3-ylbenzoic-acid
pictograms GHS07
hstatements H302+H312+H332;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart