APO424015227 Apollo Scientific Ltd

Potassium trifluoro(oxetan-3-yl)borate

  • MDL number MFCD24843091
  • Cas Number : 1430219-76-3
  • Molecular Weight : 163.98

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/potassium-trifluorooxetan-3-ylborate

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/potassium-trifluorooxetan-3-ylborate
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart