APO424015290 Apollo Scientific Ltd

(D)-(-)-2-Phenylglycinamide

  • MDL number MFCD06799064
  • Cas Number : 6485-67-2
  • Molecular Weight : 150.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 125-129
Synonym (2R)-2-Amino-2-phenylacetamide
Purity 98
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/d-2-phenylglycinamide

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/d-2-phenylglycinamide
pictograms GHS07
hstatements H317;H319
Availability details for
Quantity
  • Availability
  • Add to Cart