APO424015326 Apollo Scientific Ltd

2,2-Difluorocyclopenta-1,3-dione

  • MDL number MFCD16657105
  • Cas Number : 1215071-21-8
  • Molecular Weight : 134.08

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 72-73
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/22-difluorocyclopenta-13-dione

Safety Information

signalword Warning
pcodes P260;P280
MSDS https://store.apolloscientific.co.uk/product/22-difluorocyclopenta-13-dione
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart