APO424015341 Apollo Scientific Ltd

4-(Propan-2-yl)-1H-pyrazol-3-amine

  • MDL number MFCD11112127
  • Cas Number : 151521-49-2
  • Molecular Weight : 125.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 4-isopropyl-1H-pyrazol-3-amine
Inchi Code 1S/C6H11N3/c1-4(2)5-3-8-9-6(5)7/h3-4H,1-2H3,(H3,7,8,9)
Inchi Key UQTLFNBARWAWIS-UHFFFAOYSA-N

Description

Purity 95%
Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/4-propan-2-yl-1h-pyrazol-3-amine

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/4-propan-2-yl-1h-pyrazol-3-amine
pictograms GHS07
hstatements H302+H332;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart