APO424807114 Apollo Scientific Ltd

2,4-Dichloro-5-iodo-7H-pyrrolo[2,3-d]pyrimidine

  • MDL number MFCD13189364
  • Cas Number : 1012785-51-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Storage temp. Store at 2-8
Physical Form Solid
Shipping Temperature Ship at ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=62876

Safety Information

signalword Warning
msds http://www.apolloscientific.co.uk/display_item.php?id=62876
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart