APO424807126 Apollo Scientific Ltd

Methyl 2-chloro-5-iodobenzoate

  • MDL number MFCD07779253
  • Cas Number : 620621-48-9
  • Molecular Weight : 296.49

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/methyl-2-chloro-5-iodobenzoate

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/methyl-2-chloro-5-iodobenzoate
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart