APO424807231 Apollo Scientific Ltd

2-Amino-4-iodophenol

  • MDL number MFCD14705890
  • Cas Number : 99969-17-2
  • Molecular Weight : 235.02

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

bp 139
Iupac Name 2-amino-4-iodophenol
Inchi Code 1S/C6H6INO/c7-4-1-2-6(9)5(8)3-4/h1-3,9H,8H2
Inchi Key JRCVKCSFJNERAW-UHFFFAOYSA-N

Description

Synonym 2-Hydroxy-5-iodoaniline
Purity 95%
Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 139
Product Link https://store.apolloscientific.co.uk/product/2-amino-4-iodophenol

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/2-amino-4-iodophenol
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart