APO424807242 Apollo Scientific Ltd

3-(Bromomethyl)-5-fluorobenzoic acid

  • MDL number MFCD18378634
  • Cas Number : 1379351-68-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 3-Carboxy-5-fluorobenzyl bromide, alpha-Bromo-5-fluoro-m-toluic acid
Storage temp. Store at 2-8
Physical Form Solid
Shipping Temperature Ship at ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=63339

Safety Information

signalword Danger
msds http://www.apolloscientific.co.uk/display_item.php?id=63339
pictograms GHS05
Availability details for
Quantity
  • Availability
  • Add to Cart