APO424807474 Apollo Scientific Ltd

Methyl 3-oxetaneacetate

  • MDL number MFCD18642557
  • Cas Number : 1217800-69-5
  • Molecular Weight : 130.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Store at 2-8
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/methyl-3-oxetaneacetate

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/methyl-3-oxetaneacetate
pictograms GHS07
hstatements H302
Availability details for
Quantity
  • Availability
  • Add to Cart