APO427944237 Apollo Scientific Ltd

3-Bromo-6-chlorobenzene-1,2-diamine

  • MDL number MFCD28040528
  • Cas Number : 1823876-88-5
  • Molecular Weight : 221.48

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link https://store.apolloscientific.co.uk/product/3-bromo-6-chlorobenzene-12-diamine

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/3-bromo-6-chlorobenzene-12-diamine
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart