APO427944277 Apollo Scientific Ltd

2-(4-Chlorophenyl)-2',4'-dichloroacetophenone

  • MDL number MFCD18452152
  • Cas Number : 94171-11-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 65-66
Synonym 2-(4-Chlorophenyl)-1-(2,4-dichlorophenyl)ethanone
Storage temp. Ambient temperature
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=63899

Safety Information

signalword Danger
pcodes P260;P271;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=63899
pictograms GHS05;GHS07;GHS09
hstatements H317;H318;H411
Availability details for
Quantity
  • Availability
  • Add to Cart