APO427944281 Apollo Scientific Ltd

5-Bromo-2-fluorophenacyl bromide

  • MDL number MFCD23711739
  • Cas Number : 1427413-65-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 2-Bromo-1-(5-bromo-2-fluorophenyl)ethan-1-one
Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=63445

Safety Information

signalword Danger
pcodes P260;P271;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=63445
pictograms GHS05
hstatements H314
Availability details for
Quantity
  • Availability
  • Add to Cart